N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140-07-8
:
Formula:C₁₀H₂₄N₂O₄
MDL Number:MFCD00002854
:236.3086 g/mol
Density:1,13 g/cm³
Flash Pt:110 °C
Storage Temperature:Ambient
EINECS:205-396-0
Specification Test Results
- Appearance:Clear colorless to amber Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4990 to 1.5070 (20°C, 589 nm)
- pH:≥10 (25 % solution)
- Titration with HClO4:≥98.5 %
- Arsenic (As):≤2 ppm
- Copper (Cu):≤20 ppm
- Iron (Fe):≤400 ppm
- Lead (Pb):≤5 ppm
- Mercury (Hg):≤1 ppm
- Zinc (Zn):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.2 % (in HCl)
- Loss on ignition:≤8 % (800°C)
Specification Test Results
Frequently Bought Together