Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
6-Fluoro-3-formylchromone ≥97%
Catalog # H32857.03
CAS Number:  
Irritant
6-Fluoro-3-formylchromone ≥97%
Catalog # H32857.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.15 g/mol
Melting Pt:155…160 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:69155-76-6
Formula:C₁₀H₅FO₃
MDL Number:MFCD00139060