Accessories for LLE columns, CHROMABOND® XTR
Catalog # MANA730508
Supplier: MACHEREY-NAGEL
Specifications
- Description:PP rack, variable, for 24 positions, with 24 PP stopcocks and 24 PP needles
- Pk:1 KIT
Specifications
About this item
A variable polypropylene rack is recommended for the liquid-liquid extraction (LLE) with CHROMABOND® XTR columns.
- For application of up to 24 columns
- Flow by gravity (no vacuum)