LLE columns, CHROMABOND® XTR
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® XTR is a kieselguhr for liquid-liquid extraction (LLE).
- Coarse-grained kieselguhr (also known as diatomaceous earth, celite)
- Fast, reproducible and economical in comparison with classical LLE
- Simultaneous preparation of several samples
- No formation of emulsions
- Organic solutions need not to be dried after separation
Suitable for liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel.
Specifications
- Catalog No:
- MANA730507
- MANA730506
- MANA730505
- MANA730487.250
- MANA730509
- MANA730489
- MANA738131.150M
- MANA730502
- MANA730487
- MANA730507.100
- pH range:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13