Industrial balances, Valor™ 7000 compact bench scales
: OHAUS
A food scale designed to increase productivity of food weighing and proportioning tasks, that incorporates touchless sensors to operate certain functions without the touch of a button, thereby reducing scale contamination. Valor 7000 has Food Safety Certification (NSF) and supports a HACCP compliant quality system.
- Front and rear touchless sensors
- Dual backlit displays, front and rear for quick weighing by two operators
- One second (1 s) stabilisation time for increased throughput
- Mains supply with internal rechargeable battery for portable use
- IP 43 (splash) protection
: More connectivity options available, including optional Ethernet, second RS232 or USB. Approved versions are also available. Please see the website or enquire for more details.
Frequently Bought Together