
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1195-66-0
:
Formula:C₇H₁₅BO₃
MDL Number:MFCD03093086
:158.01 g/mol
Boiling Pt:120 °C (228 mmHg)
Density:0,966
Flash Pt:46 °C (114 °F)
Storage Temperature:Ambient
ADR:3,III
:GHS02
Related Information
Frequently Bought Together