Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
Rhodium(II) acetate dimer (∼46% Rh), anhydrous
 
undefined
 
Rhodium(II) acetate dimer (∼46% Rh), anhydrous

 

  • Appearance:
    Green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    46 % (Rh)

 

 

Frequently Bought Together