Ethyl-3-methoxypropionate for synthesis, Sigma-Aldrich®
Catalog # 8.43946.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:212-112-9
UN:3272
:GHS02
Storage Temperature:Ambient
CAS Number:10606-42-5
Formula:C₆H₁₂O₃
MDL Number:MFCD00143172