Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
 
Acetaldehyde oxime (mixture of syn and anti) for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
Acetaldehyde oxime (mixture of syn and anti) for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    185 g/l (25 °C)
  • Molar Mass:
    59.07 g/mol
  • Boiling Point:
    115 °C
  • Vapor Pressure:
    13 hPa (25 °C)
  • Flash Point:
    40 °C
  • Explosion Limit:
    4.2 - 50 %(V)
  • Density:
    0.969 g/cm3 (20 °C)
  • Ignition Point:
    380 °C

 

 

Frequently Bought Together