Order Entry
Northern Ireland
ContactUsLinkComponent
2-Mercaptopyridine-N-oxide 99%
Irritant
2-Mercaptopyridine-N-oxide 99%
Certificates
undefined
Irritant
2-Mercaptopyridine-N-oxide 99%

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Off-white to yellow to yellow-green to beige to grey-green Powder or crystals or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 72 °C
  • Titration Iodometric:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together