
4-Methoxypyridine-N-oxide ≥98%
Catalog # L17830.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:1122-96-9
:
Formula:C₆H₇NO₂
MDL Number:MFCD00006207
:125.13 g/mol
Melting Pt:99…104 °C
EINECS:214-367-1
:GHS07
Storage Temperature:Ambient