Nickel(II) tetrafluoroborate hexahydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15684-36-3
:
Formula:B₂F₈Ni·6H₂O
MDL Number:MFCD00150259
:340.394 g/mol
Density:1,5
EINECS:238-753-4
ADR:,
Related Information
Frequently Bought Together