Hexaboron silicide ≥98% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12008-29-6
:
Formula:B₆Si
MDL Number:MFCD00180907
:92.95 g/mol
Storage Temperature:Ambient
EINECS:234-535-8
Related Information
Frequently Bought Together