
3-Hydroxy-5-(trifluoromethyl)benzoic acid ≥98%
Catalog # H26157.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:328-69-8
:
Formula:C₈H₅F₃O₃
MDL Number:MFCD07368783
:206.12 g/mol
Melting Pt:191…193 °C
EINECS:None
:GHS07
Storage Temperature:Ambient