
Bis(2-methoxyethyl)phthalate ≥96%
Catalog # 44089.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
CAS Number:117-82-8
:
Formula:C₁₄H₁₈O₆
MDL Number:MFCD00042842
:282.29 g/mol
Boiling Pt:340 °C
Density:1,17
Flash Pt:185 °C (365 °F)
EINECS:204-212-6
:GHS08
Storage Temperature:Ambient