Methyl-4-formylbenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1571-08-0
:
Formula:C₉H₈O₃
MDL Number:MFCD00006950
:164.16 g/mol
Boiling Pt:264…266 °C
Melting Pt:59…63 °C
Density:1,200
Flash Pt:128 °C (262 °F)
Storage Temperature:Ambient
EINECS:216-385-5
Related Information
Frequently Bought Together