

Nickel (silver coated) (Ni:Ag; 85:15 wt%) ≥99.8% (metals basis), spheres -250 mesh
Frequently Bought Together
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Nitrile Extra Light, Nitrile Gloves
: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Magnesium ≥99%, turnings
: Thermo Fisher Scientific
Lithium hydride ≥99.4% (metals basis)
: Thermo Fisher Scientific