SPE columns, CHROMABOND® HR-P
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® HR-P is a polystyrene-divinylbenzene adsorbent resin for SPE.
- Highly porous copolymer with a very large surface
- Very high binding capacity compared with silica adsorbents
Recommended for aromatic compounds, like phenols, nitroaromatics, and pesticides from water or PAHs from oil.
Specifications
- Catalog No:
- MANA731841
- MANA731840
- MANA731839
- MANA732108
- MANA730280
- MANA730111G
- MANA730111
- MANA730108G
- 554-6594
- MANA738111.100M
- MANA730118
- MANA730117
- MANA730119
- MANA730108.250
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- Surface area:
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g
- 1200 m²/g