Respirator filters for masks, 7000 and 9000 Series, EasyLock®
Supplier: Moldex
Certificates
About this item
EasyLock® filters protect against gas, vapour and dust. Suitable for series 7000 and 9000 masks.
- Bayonet connector
- Particulate filters can either be connected with the gas cartridges or directly with the mask body
- Adaptors and pre-filters are not necessary
- Simpler logistics - 30% less parts
- Particulate filters with pleated filter technology: Significantly reduces breathing resistance, improves wearer comfort and reduces fatigue
Certifications: EN 143:2000+A1:2006 (particulate filters)
EN 14387:2004+A1:2008 (gas filters and pre-assembled filters)
EN 14387:2004+A1:2008 (gas filters and pre-assembled filters)
Frequently Bought Together