ContactUsLinkComponent
HPLC columns, organic acids analysis, NUCLEOGEL® ION 300 OA
HPLC columns, organic acids analysis, NUCLEOGEL® ION 300 OA
Supplier:  MACHEREY-NAGEL
Certificates
undefined
HPLC columns, organic acids analysis, NUCLEOGEL® ION 300 OA
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA719537
  • MANA719501
  • Max. pressure:
  • 150 bar
  • 150 bar
  • Temperature limit:
  • 95 °C
  • 95 °C
  • USP code:
  • L17
  • L17