Orders LinkContactUsLinkComponent
CA_Mainpageimage955x590.jpg
Brady Laboratory Label Recommendations
Brady offers a variety of thermal transfer label materials engineered to survive and perform in the harsh chemicals and range of temperatures of the lab environment. These durable labels remain legible and in place when put to the test.
Brady Laboratory Label Recommendations | Avantor
Brady offers a variety of thermal transfer label materials engineered to survive and perform in the harsh chemicals and range of temperatures of the lab environment. These durable labels remain legible and in place when put to the test.
Product selection
B-461image400x495.jpg
Great for flat and curved surfaces. Resistance to print smudges, solvents and extreme temperatures.
B-481image1_400x495.jpg
Designed for use in the slide staining process and other lab identification that requires superior chemical resistance.
B-488image1_400x495.jpg
Withstands numerous solvents and variable temperatures. B-488 excels at resisting chemicals, oils, alcohols and heat. 
B-490image1_400x495.jpg
Ideal for flat surfaces and can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen.
B-492image1_400x495.jpg
Engineered for wrapping around cryo vials and tubes, including already frozen vials.
B-494image1_400x495.jpg
Add color to differentiate label samples. Withstands the harshest lab enviornments including liquid nitrogen, hot water baths and more. 
B-7425image_400x495.jpg
Use on vials and tubes in applications that require resistance to chemicals and extreme temperatures. 
B-7425-ACimage1_400x495.jpg
Steam sterilization indicating polypropylene label with a preprinted header that displays "STERILIZED" when exposed to autoclave conditions
B-7425Jimage1_400x495.jpg
Ideal for full-color identification where resistance to liquid nitrogen, autoclave conditions and harsh chemicals is required.
B-403_400x495.jpg
Completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind.