ContactUsLinkComponent
Industrial scales.png
Ohaus
Ohaus offers an extended range of balances to be used in industrial environment, from compact, easy to use, for food applications (HACCP compliant) to modular models, high capacity, high level advanced with special applications, as parts counting, check weighing, and more.
weighing.png
Ranger® 7000
Simplifying even the most complex industrial applications.
  • Ten advanced application modes for complex applications, ability to control peripheral devices and/or a scale platform with optional kit
  • Fast and accurate results thanks to 1 second stabilisation time and up to 75000 d or 350000 d display resolution
  • Modular scale with IP 54 cast metal housing and sealed metal indicator to ensure durability
  • Capacity (range): 3 to 60 kg; readability (range) 0,01 to 10 g.
Product link
Brochure