Quantus Boot Covers
Features:
- Front zipper
- Elastic band in the back with adjustable press studs
- Adjustable snap fastener at instep height
- Light grey plastic sole (Maestro) high sole model (robust, waterproof, autoclavable)
- Polyester band boost between sole and texture
- Sizes: S - XL
Optional features:
- PES-ties at knee and ankle height
- Additional clip at instep height
Quantus® boot cover Plus Q-D
Features:
- Plastic zipper, concealed and offset to the side (outwards)
- Gaiter closure at top, adjusted with PES tape and buckle clip
- Two flaps as threading guide for the projecting PES tape on the edge of the gaiter
- Clip fastener at instep
- Double reinforcement on the gaiter, made of outer fabric
- Reinforcement between sole and texture, made of CONTROL fabric (white)
- Light grey nitrile plastic sole CRS-BA low (ergonomically moulded, roughened profile, stable, waterproof, autoclavable)
- Sizes: S - XL