Orders LinkContactUsLinkComponent
basan_Equip_GowningRoom.jpg
Cleanroom Furniture
Space is at a premium in cleanrooms where every square metre is expensive. As a result there are no standard solutions and no standard sizes.
Cleanroom Furniture | Avantor
The stainless steel cleanroom furniture, which is electrically conductive, consists of vertical supports with solid faces and perforated horizontal tops to reduce disturbance of the vertically directed laminar flow. Moving and cleaning the furniture is also facilitated. The holes in the tops have a diameter of 8-10 mm and are chamfered on both sides.
Product selection
basan_Equip_Chair_200.jpg
Chairs and Stools
High quality cleanroom chairs with technical and ergonomic features for health and safety. Minimal particle emission and reliable elimination of electrostatic charges.
basan_Equip_Table_200.jpg
Tables
Top solid or perforated for an optimum routing of airflow. Made from stainless steel. Equipment in chrome-plated steel, melamine face chipboard or plastic available.
basan_Equip_Shelving_200.jpg
Shelving
Cleanroom shelving made of stainless steel or chrome-plated in different heights.
basan_Equip_Cabinet_200.jpg
Cabinets
Cleanroom cabinets made of stainless steel, brushed or electropolished in many variants.
basan_Equip_Cart_200.jpg
Utility Carts
Made of stainless steel in different designs and sizes with 2 or 3 wire or solid shelves.
basan_Equip_Rack_200.jpg
Gowning Racks
Made of stainless steel. Models with compartments above the hanging rail for caps, masks etc. and with storage racks for shoes are also available.
basan_sitover_200.jpg
Sit-over Benches
Functional barrier for a controlled entry and leave of employees working in a cleanroom or in controlled environments. Cleanroom compatible sitting surfaces, optionally in polyethylene or polypropylene.
basan_Equip_Dispenser_3er_200.jpg
Dispenser Systems
Dispenser systems for storing and easy pull-out of disposables, wipers or removal of gloves from boxes.