ContactUsLinkComponent
LFKU1610-955x590.jpg
Laboratory refrigerators and freezers
Laboratory refrigerators and freezers provide safe and reliable storage for volatile or flammable substances, reagents, chemicals, poisons and samples thanks to their spark-free interiors that reduce the risk of internal explosions.
Avantor® offers a selection of safe, reliable and energy-efficient laboratory refrigerators and freezers from major brands and our own VWR® product line. Among the things to consider when purchasing a refrigerator or freezer are the size needed based on its intended location, the number of shelves needed, door style and swinging direction and available warning alarms.
HYC-118_2-540x390.jpg
Lab-grade refrigerators are important for ensuring temperature accuracy and safety.
38368635-540x390.jpg
We offer freezers in many shapes, sizes and temperature ranges, all rated highly for safety and accuracy.
Have questions about laboratory fridges and freezers?