$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
ACIDO CLORHIDRICO 37% 2.5LTS
85023-450
:
HONEYWELL INTERNATIONAL INC MX
New Product
ACIDO CLORHIDRICO 37% 2.5LTS
85023-450
:
HONEYWELL INTERNATIONAL INC MX
:
07102-2.5L-GL
New Product
Frequently Bought Together
BDH® pH Test Strips
: BDH
VWR® Traceable® Conductivity Calibration Standards (CRM)
: VWR International
Peróxido de hidrogeno 30 - 32%, ARISTAR® ULTRA, grado ultrapuro para análisis de trazas, VWR Chemicals BDH®
: BDH
Difco™ Tryptic Soy Agar, BD Biosciences
: BECTON DICKINSON DE MEXICO, S. MX