$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
SIST. CROMATOGRAFIA GASES NEXIS GC-2030
85023-154
:
INSTRUMENTACION Y SERVICIO EN MX
SIST. CROMATOGRAFIA GASES NEXIS GC-2030
85023-154
:
INSTRUMENTACION Y SERVICIO EN MX
:
221-83730-42
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH