Microcentrifuge Tubes
Screw cap microcentrifuge tube, 0.5 ml, Amber, 20000×g, Skirted, No
: 4201
Ideal for applications in freezer storage, autoclaving, boiling, or irradiation.
- External ridges fit rack serrations and allow one-handed opening
Colored PPCO screw caps are fitted with an ethylene propylene O-ring for reliable, leakproof performance. Compatible with most microcentrifuges with fixed angle heads. Withstands over 20000×g.
Tubes are made of reusable polypropylene copolymer (PPCO). Seamless conical construction with squared external threads prevents hang up of samples and reduces contamination.
: Dual-ended polypropylene tool 20170-688 provides a combination of socket wrench and bottle top action for quick, effective tube access. Tubes and caps are available separately.
- Volume:0.5 ml
- Color:Amber
- Description:Screw cap microcentrifuge tube
- Style:Skirted
- Maximum RCF:20000×g
- Graduations:No