Orders LinkContactUsLinkComponent
Microcentrifuge Tubes
Screw cap microcentrifuge tube, 0.5 ml, Amber, 20000×g, Skirted, No
  20170-723
 :  4201
Microcentrifuge Tubes
  20170-723
Microcentrifuge Tubes
Microcentrifuge Tubes
Screw cap microcentrifuge tube, 0.5 ml, Amber, 20000×g, Skirted, No
$1,744.69
Paquete de 500
Ideal for applications in freezer storage, autoclaving, boiling, or irradiation.
  • External ridges fit rack serrations and allow one-handed opening
Colored PPCO screw caps are fitted with an ethylene propylene O-ring for reliable, leakproof performance. Compatible with most microcentrifuges with fixed angle heads. Withstands over 20000×g.
Tubes are made of reusable polypropylene copolymer (PPCO). Seamless conical construction with squared external threads prevents hang up of samples and reduces contamination.
 : Dual-ended polypropylene tool 20170-688 provides a combination of socket wrench and bottle top action for quick, effective tube access. Tubes and caps are available separately.
  • Volume:
    0.5 ml
  • Color:
    Amber
  • Description:
    Screw cap microcentrifuge tube
  • Style:
    Skirted
  • Maximum RCF:
    20000×g
  • Graduations:
    No