Orders LinkContactUsLinkComponent
DECON-PHENE PLUS®, VELTEK
DECON-PHENE PLUS®, 1 gal., Concentrate, Sterile
  76628-010
 :  DPP-02
DECON-PHENE PLUS®, VELTEK
  76628-010
DECON-PHENE PLUS®, VELTEK
DECON-PHENE PLUS®, VELTEK
DECON-PHENE PLUS®, 1 gal., Concentrate, Sterile
$15,860.00
Case de 4 (1/Itm/Bx)
DECON-PHENE Plus disinfects controlled areas such as those in compounding pharmacies, biopharmaceutical, pharmaceutical, medical device, industrial facilities, diagnostic manufacturing facilities, and research laboratories. DECON-PHENE Plus is compatible with most non-porous hard surface materials and can be used on surfaces in aseptic filling and gowning rooms, in general manufacturing areas, and in laboratories.
  • Delivered with lot specific analytical, irradiation, and sterility data, tested to current USP compendium
  • Available in our convenient, one-step, ready-to-use, SimpleMix System made with USP Water for Injection (0.25 EU/mL)
  • Multiple convenient container sizes – Unit Dose, 16 oz, 1 Gallon, and 200 L either sterile or non-sterile
  • Description:
    DECON-PHENE PLUS®
  • Size:
    1 gal.
  • Sterility:
    Sterile
  • Form:
    Concentrate