Halkey-Roberts® Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 5000/Pk
MFLX06833-01BLK
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
- Housing material:Polyoxymethylene (POM) Acetal
- Descripción:Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 5000/Pk
- Environmentally Preferable:
- Pinch valve tube size:1/16" to 1/4" OD
- Garantía:None
- Referencia:MFLX06833-01BLK