Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
MFLX06290-09BLK
Provides easy single-handed disconnection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/16" ID x 5/16" ID
Construction from low-density polyethylene (LDPE).
- Tamaño de la púa/Ø int. del tubo (puerto 1):³⁄₁₆"
- Tipo de conexión:Quick-Disconnect to Barb
- Flow pattern:Straight
- Descripción:Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
- Environmentally Preferable:
- Garantía:1 year
- Referencia:MFLX06290-09BLK
- Fitting Material:LDPE
Frequently Bought Together