ContactUsLinkComponent
Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
  MFLX06290-09BLK
undefined
Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
  MFLX06290-09BLK
Provides easy single-handed disconnection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/16" ID x 5/16" ID
Construction from low-density polyethylene (LDPE).
  • Tamaño de la púa/Ø int. del tubo (puerto 1):
    ³⁄₁₆"
  • Tipo de conexión:
    Quick-Disconnect to Barb
  • Flow pattern:
    Straight
  • Descripción:
    Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
  • Environmentally Preferable:
  • Garantía:
    1 year
  • Referencia:
    MFLX06290-09BLK
  • Fitting Material:
    LDPE
Frequently Bought Together