VeriSpec® Certified Reference Material, KCl Conductivity Standards
500 ml, 100 µS/cm, Plastic bottle
Catalog # RCRV588700500A
Supplier: RICCA CHEMICAL CO
Número de proveedor #: RV588700-500A
VeriSpec® Certified Reference Material, KCl Conductivity Standards
Catalog # RCRV588700500A
Supplier: RICCA CHEMICAL CO
CAS Number:
Jump to:
Product Details & Documents
- Colorless Liquid
- Conductivity at 25 °C: See Ordering Table
Documents
Ricca Certified Reference Materials.pdf
Documentación del producto
Especificaciones
Especificaciones
- Pack type:Plastic bottle
- Size:500 ml
- Certification:ISO 17025/17034
- Conductivity:100 µS/cm
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



