Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VeriSpec® Certified Reference Material, KCl Conductivity Standards

500 ml, 10 µS/cm, Plastic bottle
Catalog # RCRV588510500A
Supplier:   RICCA CHEMICAL CO
 :  RV588510-500A
VeriSpec® Certified Reference Material, KCl Conductivity Standards
Catalog # RCRV588510500A
Supplier:  RICCA CHEMICAL CO
CAS Number:  
VeriSpec® Certified Reference Material, KCl Conductivity Standards
VeriSpec® Certified Reference Material, KCl Conductivity Standards

500 ml, 10 µS/cm, Plastic bottle

CAS Number:  
Price
$1,420.49
Log in Para disponibilidad
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Colorless Liquid
  • Conductivity at 25 °C: See Ordering Table
Especificaciones

Especificaciones

  • Pack type:
    Plastic bottle
  • Size:
    500 ml
  • Certification:
    ISO 17025/17034
  • Conductivity:
    10 µS/cm

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.