Value Plastics® Fitting, Y-Adapter, Hose Barb Union, ¹/₂" Int.Ø, Polypropylene, 50/Pack
Catálogo # MFLX40617-80BLK
Proveedor: VALUE PLASTICS TE
Acerca de este artículo
Create a secure connection in your sanitary fitting system.
- Constructed of durable PVDF
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow without particulate build-up. There are no sharp edges in the flow path to make cleaning very easy. These adapters offer good chemical resistance and may be sterilized by gamma irradiation, EtOH, or autoclave. Material meets the requirements for USP Class VI, ADCF, FDA, and are BPA-free.
- Tamaño de la púa/Ø int. del tubo (puerto 1):½"
- Tamaño de la púa/Ø int. del tubo (puerto 2):½"
- Tipo de conexión:Barbed Union
- Flow pattern:Y
- Descripción:Fitting, Y-adapter, hose barb union, ¹/₂" Int.Ø, polypropylene, 50/pack
- Environmentally Preferable:
- Garantía:Shelf life
- Referencia:MFLX40617-80BLK
- Fitting Material:PP
- Barb Size / Tubing I.D. (Port 3):½"
Frequently Bought Together