Value Plastics® Fitting, Polypropylene, Straight, Male Luer Lock to Thread, 9/16" Hex x 1/4-18; 1000/PK
Catálogo # MFLX45518-88BLK
Proveedor: VWR International
Acerca de este artículo
Easily transition between luer fittings and UNF or NPT threads.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- Tipo de conexión:Male Luer to Thread
- Flow pattern:Straight
- Descripción:Fitting, Polypropylene, Straight, Male Luer Lock to Thread, 9/16" Hex x 1/4-18; 1000/PK
- Environmentally Preferable:
- Connection type (port 1):Male luer lock
- Connection type (port 2):¹/₄-18 NPT thread
- No. of ports:2
- Referencia:MFLX45518-88BLK
- Fitting Material:PP
- Thread Size UNF(M) (Port 1):¼"-28
Frequently Bought Together