Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1" and 1 1/4" Connections
Catalog # MFLX70725-70
Supplier: VWR International
About this item
Enhance the life of your pump with service flange kits.
- Constructed of stainless steel (SS) parts
- Fits 1" and 1 1/4" connections
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
Delivery information: Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.
- Descripción:Flange Kit for 1" and 1-1/4" Connections
- Environmentally Preferable:
- Wetted materials:SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
- Garantía:1 year
- Referencia:MFLX70725-70
Frequently Bought Together