Hose Fittings, Band Clamps, for 4" OD Hose, 25/Pk
Catálogo # MFLX33350-16
Proveedor: VWR International
Acerca de este artículo
Quick and easy assembly with band clamp tool.
- Constructed of durable stainless steel
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.
- Tubo ext.Ø:101.6 mm (4")
- Descripción:Hose Fittings, Band Clamps, for 4" OD Hose, 25/Pk
- Environmentally Preferable:
- Garantía:1 year
- Referencia:MFLX33350-16