PFA Compression Female Threaded Adapter Elbows
Compression Female Threaded Adapter Elbow, PFA, ⅜" OD X ⅜" NPT(F), Compression to Thread, Elbow, ⅜", PFA
Catalog # MFLX31321-83
Supplier: VWR International
Jump to:
Product Details & Documents
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Especificaciones
- Connection Type:Compression to Thread
- Flow pattern:Elbow
- Thread size NPT(F) (port 2):⅜"
- Description:Compression Female Threaded Adapter Elbow, PFA, ⅜" OD X ⅜" NPT(F)
- Fitting Material:PFA
- Cat. No.:MFLX31321-83
- Warranty:18 months
- Sterilisation:Autoclave or ethylene oxide (ETO)