Burkle 5606-0601 Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 ml/stroke
Catalog # MFLX06043-97
Supplier: VWR International
About this item
Safely transfer aggressive chemicals.
- PTFE and FEP wetted materials withstand harsh chemicals
- Stopcock contains hazardous vapors and prevents dripping
These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).
- Velocidad de flujo:3.7 strokes/L (14 strokes/gal.)
- Descripción:Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 ml/stroke
- Environmentally Preferable:
- Wetted materials:PTFE/FEP
- Outlet connection:Discharge tubing
- Tube ext.Ø, inlet:3.2 cm (1.25")
- Tube length, inlet:60 cm (23.622")
- Tube length, outlet:120 cm (47.25")
- Garantía:15 Months
- Referencia:MFLX06043-97
Frequently Bought Together