Masterflex® Fitting, HDPE, Straight, Hose Barb Union, 3/32" ID; 100/PK
MFLX40626-44
CE certificate
Quick and easy assembly without tools.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory Approvals to match your unique application
- Ultra smooth inner surface results in fewer extractables
- Connection Type: 3/32" ID
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Tamaño de la púa/Ø int. del tubo (puerto 1):³⁄₃₂"
- Tamaño de la púa/Ø int. del tubo (puerto 2):³⁄₃₂"
- Tipo de conexión:Barbed Union
- Flow pattern:Straight
- Descripción:Fitting, HDPE, Straight, Hose Barb Union, ³⁄₃₂" ID; 100/PK
- Environmentally Preferable:
- Sterilisation:Ethylene oxide (ETO) only
- Referencia:MFLX40626-44
- Fitting Material:HDPE
Frequently Bought Together