Threaded Reducing Nipples, NPT(M)×NPT(M)
PN-2-P Threaded nipple, PFA, ⅛" male NPT Fitting, Threaded Union, Straight, ⅛", ⅛", PFA
Catalog # MFLX31320-98
CE certificate
Jump to:
Discontinued
Product Details & Documents
Create a secure connection with threaded fittings.
- Constructed of chemically inert PFA
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
Certificates
Buscar por número de loteEspecificaciones
- Connection Type:Threaded Union
- Flow pattern:Straight
- Thread size NPT(M) (port 1):⅛"
- Thread size NPT(M) (port 2):⅛"
- Description:PN-2-P Threaded nipple, PFA, ⅛" male NPT Fitting
- Fitting Material:PFA
- Cat. No.:MFLX31320-98
- Warranty:18 months
- Sterilisation:Autoclave or ethylene oxide (ETO)