Orders LinkContactUsLinkComponent
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
PN-2-P Threaded nipple, PFA, ⅛" male NPT Fitting, Threaded Union, Straight, ⅛", ⅛", PFA
Catalog # MFLX31320-98
CE certificate
Threaded Reducing Nipples, NPT(M)×NPT(M)
Catalog # MFLX31320-98
Supplier:  
CE certificate
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
PN-2-P Threaded nipple, PFA, ⅛" male NPT Fitting, Threaded Union, Straight, ⅛", ⅛", PFA
Discontinued
Product Details & Documents
Create a secure connection with threaded fittings.
  • Constructed of chemically inert PFA
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
Especificaciones
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ⅛"
  • Thread size NPT(M) (port 2):
    ⅛"
  • Description:
    PN-2-P Threaded nipple, PFA, ⅛" male NPT Fitting
  • Fitting Material:
    PFA
  • Cat. No.:
    MFLX31320-98
  • Warranty:
    18 months
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)