Order Entry
Mexico
ContactUsLinkComponent
Masterflex® Fittings, Stainless Steel, Elbow, Hose Barb Union, 3/4" ID
Masterflex® Fittings, Stainless Steel, Elbow, Hose Barb Union, 3/4" ID
Catálogo # MFLX30620-20
Proveedor:  VWR International
undefined
Masterflex® Fittings, Stainless Steel, Elbow, Hose Barb Union, 3/4" ID
Catálogo # MFLX30620-20
Proveedor:  VWR International

Acerca de este artículo

Quick and easy assembly without tools.

  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery

Create 90° bends in flexible tubing—without the kinks!

  • Tamaño de la púa/Ø int. del tubo (puerto 1):
    ¾"
  • Tamaño de la púa/Ø int. del tubo (puerto 2):
    ¾"
  • Tipo de conexión:
    Barbed Union
  • Flow pattern:
    Elbow
  • Descripción:
    Fittings, Stainless Steel, Elbow, Hose Barb Union, ¾" ID
  • Environmentally Preferable:
  • Modelo:
    30620-20
  • Orifice dimensions:
    0.6875"
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Garantía:
    1 Month
  • Referencia:
    MFLX30620-20
  • Fitting Material:
    Stainless steel 316
Frequently Bought Together