Masterflex® I/P® Standard Pump Head, Replacement End Bell Assembly, PPS Housing, SS Rotor, I/P® 73
MFLX07019-73
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Descripción:Standard Pump Head, Replacement End Bell Assembly, PPS Housing, SS Rotor, I/P® 73 Tubing
- Environmentally Preferable:
- Modelo:07019-73
- Open-head sensor:No
- Tubing sizes accepted:I/P 73
- Garantía:2 years
- Referencia:MFLX07019-73
- Rotor Material:Stainless steel
- Housing Material:PPS
Frequently Bought Together