Masterflex® Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/4" NPT(M); 10/PK
Catalog # MFLX40615-80
Supplier: VWR International
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Tamaño de la púa/Ø int. del tubo (puerto 1):0.25"
- Tamaño de la púa/Ø int. del tubo (puerto 2):¼"
- Tipo de conexión:Thread to Barb
- Flow pattern:T
- Descripción:Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/4" NPT(M); 10/PK
- Environmentally Preferable:
- Orifice dimensions:0.125"
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Referencia:MFLX40615-80
- Fitting Material:PVDF
- Barb Size / Tubing I.D. (Port 3):¼"
Frequently Bought Together