ContactUsLinkComponent
Masterflex® Quattroflow QF1200SU Disposable Pump Chamber; PP machined
  MFLX76000-02
undefined
Masterflex® Quattroflow QF1200SU Disposable Pump Chamber; PP machined
  MFLX76000-02
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
  • Ideal replacement for stainless steel single-use Quattroflow pump systems
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
  • Descripción:
    Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
  • Environmentally Preferable:
  • Garantía:
    Shelf life
  • Referencia:
    MFLX76000-02
Frequently Bought Together