ContactUsLinkComponent
Female Threaded Pipe Cap, 1/8" NPT(F), PTFE
Female Threaded Pipe Cap, 1/8" NPT(F), PTFE
Catálogo # MFLX34030-61
CE certificate
undefined
Female Threaded Pipe Cap, 1/8" NPT(F), PTFE
Catálogo # MFLX34030-61
Número de proveedor:  F-CA01
CE certificate
Acerca de este artículo
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
  • Tipo de conexión:
    Threaded Cap
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    ⅛"
  • Descripción:
    Female Threaded Pipe Cap, ⅛" NPT(F), PTFE
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Garantía:
    2 years
  • Referencia:
    MFLX34030-61
  • Fitting Material:
    PTFE
Frequently Bought Together