ContactUsLinkComponent
Connection Tubing FEP 1/4" UNF 1/16" ID×1/8" OD×1m, 1 Fitting
Connection Tubing FEP 1/4" UNF 1/16" ID×1/8" OD×1m, 1 Fitting
Catálogo # MFLX78167-55
Proveedor:  KNF NEUBERGER INC
1 year warranty
undefined
Connection Tubing FEP 1/4" UNF 1/16" ID×1/8" OD×1m, 1 Fitting
Catálogo # MFLX78167-55
Proveedor:  KNF NEUBERGER INC
Número de proveedor:  166329
1 year warranty
Acerca de este artículo
Replacement parts keep your dosing pump running smoothly.
  • Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
  • Descripción:
    Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
  • Environmentally Preferable:
  • Wetted materials:
    FEP
  • Garantía:
    1 year
  • Referencia:
    MFLX78167-55
Frequently Bought Together