Orders LinkContactUsLinkComponent
Masterflex® B/T® Replacement Fuses, Avantor®
Replacement Fuse, 3,15 A/250 V, 5 X 20 mm, Slo-Blo, Masterflex L/S and I/P drives
Catalog # MFLX77500-25
 :  77500-25
Masterflex® B/T® Replacement Fuses, Avantor®
Catalog # MFLX77500-25
Masterflex® B/T® Replacement Fuses, Avantor®
Masterflex® B/T® Replacement Fuses, Avantor®
Replacement Fuse, 3,15 A/250 V, 5 X 20 mm, Slo-Blo, Masterflex L/S and I/P drives
Price
$779.00
Log in Para disponibilidad
Each (2UN) Paquete de 2
Product Details & Documents
Make the most of your drive with these replacement parts.
  • Optimize performance of your Masterflex pumps
Suitable for use with Masterflex® B/T® drive 77111-30.
Especificaciones
  • Description:
    Replacement Fuse, 3,15 A/250 V, 5 X 20 mm, Slo-Blo
  • Compatible with:
    Masterflex L/S and I/P drives