Masterflex® L/S® High-Performance Pump Head Mounting Hardware, Avantor®
L/S® High-Performance Pump Head Replacement Tubing Retainer Kit for High-Performance Precision Tubing. 2 years warranty, L/S
MFLX77252-64
: 77252-64
Masterflex® L/S® High-Performance Pump Head Mounting Hardware, Avantor®
MFLX77252-64
Secure your Masterflex® High-Performance pump head!
- Constructed of durable stainless steel
Keep your pump head operating at its best with these replacement parts.
: Includes four 10-24 stainless steel mounting screws.
- Description:L/S® High-Performance Pump Head Replacement Tubing Retainer Kit for High-Performance Precision Tubing. 2 years warranty
- Pump Series:L/S