Masterflex® L/S® Easy-Load® II 2-Channel Pump Head Tubing Retainer Kit for Precision Tubing
Catálogo # MFLX77200-94
Proveedor: Avantor Fluid Handling
Acerca de este artículo
Enhance the life of your pump head with the replacement retainer kits.
- Retainer kits provide the parts necessary to replace the spring-loaded tubing retainers in your Masterflex® Easy-Load® II pump heads
- Suitable for use with Easy-Load® II pump heads 77202-50 and -60
Replacement parts to keep your pump head operating even longer. Tubing retainers hold the tubing in place within the pump head. Pump head sold separately.
Entrega: Includes two small upper retainers, two small lower retainers, and two compression springs.
- Descripción:Replacement Tubing Retainer Kit for L/S Easy-Load II Pump Heads 77202-50 and -60
- Environmentally Preferable:
- Garantía:1 year
- Referencia:MFLX77200-94
Frequently Bought Together